This product will no longer be available on thermofisher.com after Dec. 28, 2020. Learn more »
Thermo Scientific™
Richard-Allan Scientific™ Papanicolaou Stain EAs
Related applications:
Anatomical Pathology
Control the color hues and intensities of your work with Thermo Scientific™ Richard-Allan Scientific™ Papanicolaou Stain EAs, useful for both gynecological and non-gynecological staining.
Back to top
  • EA-36 and EA-50 are used in conjunction with OG-6 for gynecological staining.
  • EA-65 is used with OG-6 for non-gynecological staining.
  • Modified EA is a Gill modification that minimizes precipitation, reduces staining time, and does not contain Bismarck Brown.