Yeast beta-Galactosidase Assay Kit
Thermo Scientific™

Yeast beta-Galactosidase Assay Kit

Extract protein from yeast two-hybrid reporter systems and measure beta-galactosidase activity using compatible Y-PER Reagent and b-Gal Assay Reagent.

Have Questions?
Catalog NumberQuantity
7576875 mL
Catalog number 75768
Price (USD)
304.00
Each
Quantity:
75 mL
Request bulk or custom format
Price (USD)
304.00
Each

Thermo Scientific Yeast β-Galactosidase Assay Kit is specially formulated for yeast cells and ideal for identifying protein interactions in-vivo using two-hybrid systems.

Features of Yeast beta-Galactosidase Assay Kit:

  • Efficient lysis of yeast cells and a colorimetric detection system
  • Quantitative or qualitative assay for yeast colonies grown on solid media or in suspension
  • Colorimetric assay easily measured in a plate reader or spectrophotometer (420nm)
  • Adaptable to 96-well microplates or tube assay formats
  • Allows cells obtained from solid media or liquid culture to be assayed directly with no harvesting or washing steps.
  • Perform homogeneous screening assays or measure activity secondarily after cell lysis
  • Also compatible with prepared lysates from other species such as bacteria

Yeast β-Galactosidase Assay Kit allows qualitative or quantitative determination of β-galactosidase activity in solution directly from colonies growing on solid medium. A portion of the colony is suspended in a mixture of Thermo Scientific Y-PER Yeast Protein Extraction Reagent and β-Galactosidase Assay Buffer. After a brief incubation period, the solution turns yellow from the hydrolysis of o-nitrophenyl-β-D-galactopyranoside (ONPG) to o-nitrophenol (ONP) and galactose in a mildly alkaline solution. The assay becomes quantitative if calibrated to cell number by measuring the optical density at 660nm.

The gene encoding β-galactosidase (lacZ) of E. coli has been widely used as a reporter gene in many different prokaryotic and eukaryotic organisms. β-Galactosidase activity is also commonly used as an indicator of protein-protein interactions in-vivo using two-hybrid systems. The interaction strength is verified or quantitated using a β-galactosidase activity assay.

For Research Use Only. Not for use in diagnostic procedures.

The gene encoding β-galactosidase (lacZ) of E. coli has been widely used as a reporter gene in many different prokaryotic and eukaryotic organisms. β-Galactosidase activity is also commonly used as an indicator of protein:protein interactions in vivo using two-hybrid systems. The interaction strength is verified or quantitated using a β-galactosidase activity assay.

Common protocols for determining β-galactosidase activity from liquid cultures involve harvesting cells by centrifugation, washing cells several times, and lysing cells by either subjecting them to freeze/thaw cycles or by treating them with detergents or organic solvents. These traditional protocols are not amenable to projects that involve screening cells growing in 96-well plates. The Yeast β-Galactosidase Assay Kit allows cells obtained from solid media or liquid culture to be assayed directly with no harvesting or washing steps.

Specifications
Detection MethodAbsorbance
Dye TypeEnzyme Labeled
Format96-well Plate
Quantity75 mL
SubstrateONPG
Substrate TypeBeta-Gal Substrate
Sufficient For250 colonies or 250 culture samples (100 μL each)
TargetBeta-Galactosidase
Absorbance Wavelength420 nm
For Use With (Application)Absorbance
For Use With (Equipment)Microplate Reader
Product LinePierce
Product TypeBeta-Galactosidase Assay Kit
Reference Wavelength660 nm
Unit SizeEach
Contents & Storage
Sufficient For: 250 colonies or 250 culture samples (100 μL each)
  • Y-PER Yeast Protein Extraction Reagent, 25 mL (store at room temperature)
  • 2X beta-Galactosidase Assay Buffer, 25 mL (store at -20°C)
  • 1M Sodium Carbonate Stop Solution, 25 mL (store at room temperature)