Yeast beta-Galactosidase Assay Kit

Catalog number: 75768B

Thermo Scientific™  Related applications: Protein Assays and Analysis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Thermo Scientific Yeast β-Galactosidase Assay Kit is specially formulated for yeast cells and ideal for identifying protein interactions in-vivo using two-hybrid systems.

Features of Yeast beta-Galactosidase Assay Kit:

• Efficient lysis of yeast cells and a colorimetric detection system
• Quantitative or qualitative assay for yeast colonies grown on solid media or in suspension
• Colorimetric assay easily measured in a plate reader or spectrophotometer (420nm)
• Adaptable to 96-well microplates or tube assay formats
• Allows cells obtained from solid media or liquid culture to be assayed directly with no harvesting or washing steps.• Perform homogeneous screening assays or measure activity secondarily after cell lysis
• Also compatible with prepared lysates from other species such as bacteria

Yeast β-Galactosidase Assay Kit allows qualitative or quantitative determination of β-galactosidase activity in solution directly from colonies growing on solid medium. A portion of the colony is suspended in a mixture of Thermo Scientific Y-PER Yeast Protein Extraction Reagent and β-Galactosidase Assay Buffer. After a brief incubation period, the solution turns yellow from the hydrolysis of o-nitrophenyl-β-D-galactopyranoside (ONPG) to o-nitrophenol (ONP) and galactose in a mildly alkaline solution. The assay becomes quantitative if calibrated to cell number by measuring the optical density at 660nm.

The gene encoding β-galactosidase (lacZ) of E. coli has been widely used as a reporter gene in many different prokaryotic and eukaryotic organisms. β-Galactosidase activity is also commonly used as an indicator of protein-protein interactions in-vivo using two-hybrid systems. The interaction strength is verified or quantitated using a β-galactosidase activity assay.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Assay Type: Reporter Enzyme
Compatible Cells: Yeast Cells
Detection Method: Colorimetric
For Use With (Equipment): Microplate Reader
Format: 96-well plate
Product Line: Pierce™
Product Size: Custom
Substrate: ONPG
Substrate Properties: Chemical Substrate
Substrate Type: Beta-Gal Substrate
Target Enzyme: Beta-Galactosidase
Technique: Absorbance

Contents & storage

• Y-PER™ Yeast Protein Extraction Reagent; store at room temperature
• 2X β-Galactosidase Assay Buffer; store at -20°C
• β-Galactosidase Assay Stop Solution: store at room temperature

Documents

Manuals & protocols