Regulatory requirements vary by country; products may not be available in your geographic area.
TMB
Catalog number: 7588925
 Related applications: Animal Health
Error loading your content!
  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons
Your on-site stock
›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top
Description
TMB (3,3´,5,5´-tetramethylbenzidine) is the most popular chromogenic substrate for horseradish peroxidise (HRP) detection in ELISA, yielding a blue color when oxidized.
For Veterinary Use Only.
Specifications
Product Line: Prionics™
Product Size: 60 mL
Substrate: TMB
Substrate Type: HRP (Horseradish Peroxidase) Substrate
Documents
Manuals & protocols