Regulatory requirements vary by country; products may not be available in your geographic area.
PrioCHECK™ PRV gB Antibody ELISA Kit
Applied Biosystems™

PrioCHECK™ PRV gB Antibody ELISA Kit

Have Questions?
Catalog NumberQuantity
7589100450 samples
Catalog number 7589100
Price (USD)
-
Quantity:
450 samples
The PrioCHECK® PRV gB ELISA is a blocking ELISA that detects antibodies against the Pseudorabies virus (PRV) glycoprotein B in pig serum samples. The samples are incubated in a microtiter plate coated with an inactivated PRV antigen. After washing, the plates are incubated with a monoclonal antibody (mAb) directed against an epitope located on glycoprotein B (gB) of the PRV. This mAb is pre-labeled with an enzyme that generates a color signal. The signal is measured and when no color is formed the sample contains anti-PRV antibodies that competed for the PRV antigen. With this result, the sample is positive for PRV.
Specifications
Assay SensitivitySee product documentation
Product LinePrioCHECK, Prionics
Protein FamilyOther Proteins
Quantity450 samples
Target Kit NamedPRV gB Antibody
Target Organism ClassAujeszkyins Disease (Pseudorabies Virus)
Target Species ValidatedPorcine
Immunoassay Kit FormatOther Format
Target SpeciesPorcine
Unit SizeEach
Contents & Storage
5 plates