Regulatory requirements vary by country; products may not be available in your geographic area.

Prion Monoclonal Antibody 6H4, POD-conjugate

Catalog number: 7610001

 Related applications: Animal Health

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Anti-prion 6H4 labeled with horse radish peroxidase (6H4-POD). Mouse IgG1 subtype. Light chain: k subtype. mAb 6H4 recognizes the sequence DYEDRYYRE of the prion protein (human PrP: amino acids 144-152). The antibody reacts with most known mammalian PrP sequences (human, cattle, sheep, rabbit, mink, a variety of primates, mouse, hamster, and rat).
For Veterinary Use Only.

Specifications

Clonality: Monoclonal
Host Species: Mouse
Product Line: Prionics™
Product Size: 100 µg
Reactivity: Bovine, Hamster, Mink, Mouse, Primate, Rabbit, Rat, Sheep

Documents

Manuals & protocols