Regulatory requirements vary by country; products may not be available in your geographic area.

PrioCON CSFV Monoclonal Antibody 21.2 (A1 domain of E2 protein), HRPO-labeled

Catalog number: 7610010

 Related applications: Animal Health

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Monoclonal antibody conjugate raised against the A1 domain of CSFV E2 protein that is present in all CSFV field strains. Used for immuno-peroxidase test on cryostat sections. Working dilution is 1:100. One vial is sufficient for 650 cryostat sections.
For Veterinary Use Only.

Specifications

Clonality: Monoclonal
Host Species: Mouse
Product Line: PrioCON, Prionics™
Product Size: 1 mL
Reactivity: Swine

Documents

Manuals & protocols