Regulatory requirements vary by country; products may not be available in your geographic area.

PrioCON CSFV Monoclonal Antibody 39.5 (A1 domain of E2 protein), HRPO-labeled

Catalog number: 7610015

 Related applications: Animal Health

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The monoclonal antibody conjugate recognizes the A1 domain of the CSFV E2 protein. Can be used to directly stain non-neutralized CSFV in microplate neutralization assays. Working dilution is 1:100. One vial is sufficient for 20 96-well plates.
For Veterinary Use Only.

Specifications

Clonality: Monoclonal
Host Species: Mouse
Product Size: 1 mL
Reactivity: Swine

Documents

Manuals & protocols