DNA Isolation Kit
Catalog number:  761001D
Error loading your content!
  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons
Your on-site stock
›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top
Description
A modified salting-out technique used to isolate DNA from whole blood and buffy coat samples. Produces a high yield of DNA. Application: High-quality DNA can be used for PCR and other downstream applications. DNA is stable for long-term storage
Contents & storage
Store at room temperature.
Documents
Manuals & protocols