Regulatory requirements vary by country; products may not be available in your geographic area.

PrioCON CSFV Monoclonal Antibody 44.3 (C domain of E2 protein), HRPO-labeled

Catalog number: 7610020

 Related applications: Animal Health

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Monoclonal antibody conjugate raised against the C domain of CSFV E2 protein. The C domain is absent in some strains of CSFV. Can be used in munoperoxidase test on cryostatic sections. Working dilution is 1:100. One vial is sufficient for 650 cryostat sections.
For Veterinary Use Only.

Specifications

Clonality: Monoclonal
Host Species: Mouse
Product Line: PrioCON, Prionics™
Product Size: 1 mL
Reactivity: Swine

Documents

Manuals & protocols