PrioCHECK™ Bovine BVDV PIfocus Ag Strip Kit
Applied Biosystems™

PrioCHECK™ Bovine BVDV PIfocus Ag Strip Kit

Note: The name of this product has changed. It was previously called PRIOCHECK BVDV AG PI FOCUS. The PrioCHECK™ BVDVRead more
Have Questions?
Catalog NumberQuantity
7610140900 samples, 10 plates
Catalog number 7610140
Price (EUR)
-
Quantity:
900 samples, 10 plates
Note: The name of this product has changed. It was previously called PRIOCHECK BVDV AG PI FOCUS.

The PrioCHECK™ BVDV Antigen PIfocus kit is a diagnostic solution, based on ELISA technology, that detects persistently infected cattle with high accuracy. The ELISA can detect the BVDV-antigen in ear tissue samples and can differentiate between persistently infected (PI) and acutely infected animals. The PrioCHECK™ BVDV Ag PIfocus assay is a rapid, sensitive, and highly specific test for the detection of PI-animals that demonstrate the BVDV p80 antigen. The method for BVDV antigen detection is a sandwich ELISA giving a signal proportional to the sample antigen concentration. The antigen test detects the BVDV using two monoclonal antibodies (mAbs) that recognize different epitopes on the highly conserved NS3 non-structural protein (p80) of BVDV. One of these mAbs is labelled with an enzyme that generates a color signal. The sample is positive when color has formed.
Specifications
Assay SensitivitySee product documentation
Gene ID (Entrez)Non-Gene
Gene SymbolNon-Gene
Immunoassay Kit GradeStrip
Product LinePrioCHECK, Prionics
Protein FamilyOther Proteins
Quantity900 samples, 10 plates
Target Kit NamedBVDV Antigen Plfocus
Target Organism ClassBovine Viral Diarrhea Virus
Target Species ValidatedBovine
Immunoassay Kit FormatOther Format
Target SpeciesBovine
Unit SizeEach
Contents & Storage
10 Plates