Regulatory requirements vary by country; products may not be available in your geographic area.
PrioCHECK™ Porcine SVDV Ab Kit
Applied Biosystems™

PrioCHECK™ Porcine SVDV Ab Kit

Note: The name of this product has changed. It was previously called PRIOCHECK SVDV AB ELISA. The PrioCHECK™ SVDV AntibodyRead more
Have Questions?
Catalog NumberQuantity
7610205440 samples, 5 plates
Catalog number 7610205
Price (USD)
-
Quantity:
440 samples, 5 plates
Note: The name of this product has changed. It was previously called PRIOCHECK SVDV AB ELISA.

The PrioCHECK™ SVDV Antibody ELISA is an established test that has been successfully applied in the Dutch National Screening program for SVDV. It is the test of choice for the control of SVDV outbreaks in many European countries because of its proven high specificity and sensitivity. The sera are tested at a single dilution (1:5), incubations are carried out at room temperature, and test results are available within 3 hours. Together, these factors make the PrioCHECK™ SVDV Antibody ELISA simple, easy to automate, and therefore suitable for screening large numbers of serum samples.

Test procedure
The PrioCHECK™ SVDV Antibody ELISA is a competitive double antibody sandwich ELISA for the detection of specific antibodies against the Swine Vesicular Disease virus (SVDV) found in the serum and plasma of pigs. Plates are coated with anti-SVDV specific monoclonal antibodies to which an inactivated SVDV antigen is bound. The sample and the detection antibody (a monoclonal antibody labeled with an enzyme that generates a color signal) are then added. The SVDV-specific antibodies present in the test sample and the detection antibody compete for binding to the SVDV antigen. The signal is then measured and when no color is formed, antibodies in the sample have competed for the available viral protein. With this result, the sample is positive for SVDV.
Specifications
Assay SensitivitySee product documentation
Gene ID (Entrez)Non-Gene
Gene SymbolNon-Gene
Immunoassay Kit GradeStrip
Product LinePrioCHECK, Prionics
Protein FamilyOther Proteins
Quantity440 samples, 5 plates
Target Kit NamedSVDV Antibody
Target Organism ClassSwine Vesicular Disease Virus (SVDV)
Target Species ValidatedPorcine
Immunoassay Kit FormatOther Format
Target SpeciesPorcine
Unit SizeEach
Contents & Storage
5 Plates