PrioCHECK™ Porcine Toxoplasma Ab Kit
Applied Biosystems™

PrioCHECK™ Porcine Toxoplasma Ab Kit

Note: The name of this product has changed. It was previously called PRIOCHECK TOXOPL AB PORCIN. The PrioCHECK™ Toxoplasma AntibodyMás información
Have Questions?
Número de catálogoCantidad
76102305 placas
Número de catálogo 7610230
Precio (MXN)
-
Cantidad:
5 placas
Note: The name of this product has changed. It was previously called PRIOCHECK TOXOPL AB PORCIN.

The PrioCHECK™ Toxoplasma Antibody ELISA is a reliable and efficient diagnostic test for the detection of antibodies against Toxoplasma gondii in porcine serum, plasma, and meat juice samples and can be used for monitoring and surveillance purposes.The same sample used in the PrioCHECK™ Toxoplasma Antibody ELISA can be also tested with other PrioCHECK™ diagnostic products for various other pig diseases including Trichinella and Salmonella infections. This makes the test an excellent and valuable tool for porcine food safety programs. The PrioCHECK™ Toxoplasma Antibody ELISA is also easy to use. The kit contains colored buffers, helping to reduce handling errors.

Test procedure
The PrioCHECK™ Toxoplasma Antibody ELISA is an indirect ELISA for the detection of antibodies against Toxoplasma gondii. The test follows a short four-step ELISA protocol. Ninety prepared samples can be analyzed within 150 minutes. Test samples are incubated in plates coated with Toxoplasma antigen at room temperature. Plates are then washed and an enzyme labeled anti-pig antibody is added. The signal is measured and if color develops the sample is positive for anti-Toxoplasma antibodies.
Especificaciones
Sensibilidad del ensayoSee product documentation
ID de gen (Entrez)Sin genes
Símbolo de genSin genes
Grado de kit de inmunoensayoStrip
Línea de productosPrioCHECK, Prionics
Familia de proteínasOtras proteínas
Cantidad5 placas
Nombre del kit de destinoToxoplasma Antibody
Clase de organismo dianaToxoplasma gondii
Especies diana validadasPorcine
Formato del kit de inmunoensayoOther Format
Especies dianaPorcine
Unit SizeEach
Contenido y almacenamiento
5 Plates

Preguntas frecuentes

Are negative PP values (see INSTRUCTIONS FOR USE here: https://assets.thermofisher.com/TFS-Assets/LSG/manuals/MAN0013858_7610230_UG_en.pdf) acceptable for the PrioCHECK Porcine Toxoplasma Ab Kit (Cat. No. 7610230)?

Yes, negative PP values are normal for this kit. In the QC panel for this kit's release, there were serum samples that gave negative % PP results.

Find additional tips, troubleshooting help, and resources within our Animal Health Support Center.