Regulatory requirements vary by country; products may not be available in your geographic area.

PrioCHECK® Toxoplasma Antibody ELISA Kit, porcine, strip

Catalog number: 7610270

 Related applications: Animal Health

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The PrioCHECK® Toxoplasma Antibody ELISA is a reliable and efficient diagnostic test for the detection of antibodies against Toxoplasma gondii in porcine serum, plasma, and meat juice samples and can be used for monitoring and surveillance purposes.The same sample used in the PrioCHECK® Toxoplasma Antibody ELISA can be also tested with other PrioCHECK® diagnostic products for various other pig diseases including Trichinella and Salmonella infections. This makes the test an excellent and valuable tool for porcine food safety programs. The PrioCHECK® Toxoplasma Antibody ELISA is also easy to use. The kit contains colored buffers, helping to reduce handling errors.

Test procedure
The PrioCHECK® Toxoplasma Antibody ELISA is an indirect ELISA for the detection of antibodies against Toxoplasma gondii. The test follows a short four-step ELISA protocol. Ninety prepared samples can be analyzed within 150 minutes. Test samples are incubated in plates coated with Toxoplasma antigen at room temperature. Plates are then washed and an enzyme labeled anti-pig antibody is added. The signal is measured and if color develops the sample is positive for anti-Toxoplasma antibodies.
For Veterinary Use Only.

Specifications

Protein Family: Other Proteins
Assay Sensitivity: See product documentation
Gene ID: Non-Gene
Gene Symbol: Non-Gene
Immunoassay Kit Format: Various Other
Immunoassay Kit Grade: Strip
Product Line: PrioCHECK®, Prionics™
Product Size: 4500 samples
Target (Kit Named): Toxoplasma Antibody
Target Organism Class: Toxoplasma gondii
Target Species (Named): Porcine
Target Species (Validated): Porcine

Contents & storage

50 plates

Documents

Manuals & protocols