Regulatory requirements vary by country; products may not be available in your geographic area.

PrioCHECK® Trichinella Antibody ELISA Kit, strip

Catalog number: 7610280

 Related applications: Animal Health

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The PrioCHECK® Trichinella Antibody ELISA is a highly sensitive and specific ELISA for the detection of antibodies against Trichinella in serum and meat juice samples of pigs. No cross reactivity with commonly found pig parasites has been observed.

Test procedure
The PrioCHECK® Trichinella Antibody ELISA is a diagnostic kit based on ELISA technology. It follows a four-step protocol consisting of sample preparation, sample incubation, conjugate incubation, and detection. Serum or meat juice samples are incubated in an ELISA plate coated with the Trichinella E/S antigen. Anti-Trichinella antibodies present in the sample bind to the antigen. Plates are then washed and an anti-swine antibody labeled with horse radish peroxidase (HRP) is used to detect antibodies bound to the E/S antigen. Color development using mittels tetramethyl-benzidine (TMB) substrate measured optically at a wavelength of 450 nm shows the presence of antibodies directed against Trichinella spp.
For Veterinary Use Only.

Specifications

Assay Sensitivity: See product documentation
Gene ID: Non-Gene
Gene Symbol: Non-Gene
Immunoassay Kit Format: Various Other
Immunoassay Kit Grade: Strip
Product Line: PrioCHECK®, Prionics™
Product Size: 4500 samples
Target (Kit Named): Trichinella Antibody
Target Organism Class: Trichinella
Target Species (Named): Porcine
Target Species (Validated): Porcine

Contents & storage

50 plates

Documents

Manuals & protocols