Regulatory requirements vary by country; products may not be available in your geographic area.

PrioCHECK® Brucella Antibody ELISA Kit, strip

Catalog number: 7610400

 Related applications: Animal Health

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The PrioCHECK® Brucella Antibody ELISA is an indirect ELISA for the detection of antibodies against Brucella abortus and Brucella melitensis in the serum and milk of cattle, sheep, and goats. The PrioCHECK® Brucella Antibody ELISA procedure consists of 4 steps. In the first step, serum samples and milk samples are dispensed in the coated wells of a microtiterplate. Antibodies, directed against Brucella abortus and Brucella melitensis, present in the test sample, will bind to the antigen during incubation. The bound antibodies are detected using an anti-Ig monoclonal antibody, conjugated to an enzyme that generates a color signal. Color development will occur when specific antibodies against Brucella abortus or Brucella melitensis are present in the test sample. Color development indicates the presence of Brucella antibodies in the sample.
For Veterinary Use Only.

Specifications

Protein Family: Other Proteins
Assay Sensitivity: See product documentation
Gene ID: Non-Gene
Gene Symbol: Non-Gene
Immunoassay Kit Format: Various Other
Immunoassay Kit Grade: Strip
Product Line: PrioCHECK®, Prionics™
Product Size: 460 samples
Target (Kit Named): Brucella Antibody
Target Organism Class: Brucella
Target Species (Named): Bovine
Target Species (Validated): Bovine, Goat, Sheep

Contents & storage

5 plates

Documents

Manuals & protocols