Regulatory requirements vary by country; products may not be available in your geographic area.
PrioCHECK™ Porcine Salmonella Kit
Applied Biosystems™

PrioCHECK™ Porcine Salmonella Kit

Note: The name of this product has changed. It was previously called PRIOCHECK SALMONELLA AB PORCINE. The PrioCHECK™ Salmonella AntibodyRead more
Have Questions?
Catalog NumberQuantity
76104804500 samples, 50 plates
Catalog number 7610480
Price (USD)
-
Quantity:
4500 samples, 50 plates
Note: The name of this product has changed. It was previously called PRIOCHECK SALMONELLA AB PORCINE.

The PrioCHECK™ Salmonella Antibody ELISA is a fast, effective, and economical test for large-scale screening of pigs and for application in control programs of Salmonella infections. The PrioCHECK™ Salmonella Antibody ELISA uses purified lipopolysaccharide (LPS) O-antigen of S. Typhimurium and S. Choleraesuis. This makes the PrioCHECK™ Salmonella Antibody ELISA specific for the serogroup B, C1, and D (the most common serotypes in Europe, Asia, and America).

Test procedure
The PrioCHECK™ Salmonella Antibody ELISA is an indirect ELISA for the detection of antibodies against Salmonella LPS O-antigens 1, 4, 5, 6, 7, and 12 found in pig serum, plasma, and meat juice samples. Test samples are incubated in plates coated with the purified LPS isolated from S. Typhimurium and S. Choleraesuis at room temperature. Plates are then washed and an anti-swine antibody labeled with an enzyme that generates a color signal is added. The signal is measured and if color develops the sample is positive for anti-Salmonella LPS antibodies. Color intensity is directly related to the amount of anti-Salmonella antibody present in the test sample.
Specifications
Assay SensitivitySee product documentation
Gene ID (Entrez)Non-Gene
Gene SymbolNon-Gene
Immunoassay Kit GradeStrip
Product LinePrioCHECK, Prionics
Protein FamilyOther Proteins
Quantity4500 samples, 50 plates
Target Kit NamedSalmonella Antibody
Target Organism ClassSalmonella
Target Species ValidatedPorcine
Immunoassay Kit FormatOther Format
Target SpeciesPorcine
Unit SizeEach
Contents & Storage
50 Plates