PrioCHECK™ FMDV Type Asia1 Antibody ELISA Kit, strip
Applied Biosystems™

PrioCHECK™ FMDV Type Asia1 Antibody ELISA Kit, strip

The PrioCHECK™ family of type-specific FMD tests consists of ELISA test kits for the detection of serological antibodies directed against specific FMDV structural protein compositions derived from FMDV-O, FMDV-A, or FMDV-Asia1 virus strains.
Have Questions?
Catalog NumberQuantity
7610870450 samples
Catalog number 7610870
Price (MXN)
-
Quantity:
450 samples

The PrioCHECK™ family of type-specific FMD tests consists of ELISA test kits for the detection of serological antibodies directed against specific FMDV structural protein compositions derived from FMDV-O, FMDV-A, or FMDV-Asia1 virus strains.

Test procedure
In the PrioCHECK™ FMDV Type Asia1 ELISA, plates are coated with the FMDV type Asia1 antigen. The key reagent of the test is an anti-FMDV specific monoclonal antibody (mAb) conjugated to an enzyme that generates a color signal. Binding of the conjugate to the immobilized antigen is blocked by anti-FMDV antibodies present in the test sample. The signal is then measured and when no color is formed, the antibodies in the sample have competed for the viral protein coated on the plate. Samples with a percent inhibition (PI) value of ≥ 50% have shown an immune response to the foot and mouth disease virus type Asia 1. With this result, the sample is immuno-reactive (i.e., anti-FMDV antibodies are present).

Specifications
Assay SensitivitySee product documentation
Gene ID (Entrez)Non-Gene
Gene SymbolNon-Gene
Immunoassay Kit GradeStrip
Product LinePrioCHECK, Prionics
Product TypeELISA Kit
Protein FamilyOther Proteins
Quantity450 samples
Target Kit NamedFMDV Type Asia1 Antibody
Target Organism ClassFoot and Mouth Disease Virus (FMDV)
Target Species ValidatedAll Sensitive Animals
Immunoassay Kit FormatOther Format
Target SpeciesLivestock
Unit SizeEach
Contents & Storage
5 plates