Thermo Scientific™
Richard-Allan Scientific™ Mucolytic Agent
 Related applications:
Anatomical Pathology
Easily identify specimens through the tint of Thermo Scientific™ Richard-Allan Scientific™ Mucolytic Agent.
Error loading your content!
  Catalog number Price ({{currency}}) Availability Qty
{{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}} Add To Cart
{{addItemToList.successMessage}} {{addItemToList.listName}}
Back to top
Description
  • Preserve and liquefy mucoid cytology specimens to optimize diagnostic results
  • Maintain cell morphology, size, and staining characteristics of fresh and unfixed cytology specimens
  • Store specimens indefinitely with no adverse effects
Documents
Manuals & protocols