Regulatory requirements vary by country; products may not be available in your geographic area.

PrioCHECK® Brucella Rose Bengal Antigen

Catalog number: 7620301

 Related applications: Animal Health

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Brucella Rose Bengal Antigen is a stained, killed, and standardized suspension of Brucella abortus bacteria supplied in a convenient dropper bottle for ease of use. The reagent is used in the rapid agglutination test. This test depends on the ability of antibody in the serum to agglutinate the stained bacterial antigen. When this occurs, the aggregates become clearly visible to the naked eye. The presence or absence of a visible agglutination indicates the presence or absence of antibodies in the samples tested.
For Veterinary Use Only.

Specifications

Target Organism Class: Brucella
Validated Application: Agglutination Test
Detection Method: Agglutination
Disease: Brucellosis
Product Line: PrioCHECK®, Prionics™
Product Size: 10 mL
Species: Cattle⁄Bovine, Goat⁄Caprine, Sheep⁄Ovine

Documents

Manuals & protocols