Custom, bulk or OEM requests for this product—Complete this form to receive more information.
Product Image
Invitrogen™

Poly(A) Tail-Length Assay Kit

This kit utilizes a novel G/I extension technique which preserves the 3' end of RNAs and ensures accurate measurement of poly(A) tail-length. And, all reaction components have been optimized so you get quality data quickly.
Have Questions?
Catalog NumberQuantity
764551KT1 kit
Catalog number 764551KT
Price (USD)
806.65
Online exclusive
906.00
Save 99.35
Each
Quantity:
1 kit
Price (USD)
806.65
Online exclusive
906.00
Save 99.35
Each
Poly adenylation of eukaryotic mRNAs plays a critical role in RNA metabolism, including stability, enhanced translation, nuclear export, and miRNA mediated gene regulation. The Poly(A) Tail-Length Assay Kit allows you to quickly measure the poly(A) tails of multiple mRNAs in only three hours. This kit utilizes a novel G/I extension technique that preserves the 3' end of RNAs and ensures accurate measurement of poly(A) tail-length. And all reaction components have been optimized so you get quality data quickly.

Features

  • Measure the poly(A) tail of multiple RNAs in under 3 hrs using common lab equipment
  • Accurate measurement of poly(A) tail length compared to other assays
  • One kit includes all necessary reagents for 5 G/I tailing, 20 RT, and 80 PCR reactions

Technology

The Poly(A) Tail-Length Assay Kit uses four key steps to enable poly(A) tail-length determination (see figure).

  • Step 1—poly(A) polymerase adds a limited number of guanosine and inosine residues to the 3'-ends of poly(A)-containing RNAs.
  • Step 2—the tailed-RNAs are converted to DNA through reverse transcription using the newly added G/I tails as the priming sites.
  • Step 3—PCR amplification products are generated using two primer sets. A gene-specific forward and reverse primer set designed upstream of the polyadenylation site (e.g., the 3'-UTR) is produced as a control for the gene-of-interest. The second set of primers uses the gene-specific forward primer and the universal reverse primer provided with the kit to generate a product that includes the poly(A) tails of the gene-of-interest.
  • Step 4—the PCR products are separated on an agarose or polyacrylamide gel (see figure).

Selected references

  • Parker, R. and Song, H. (2004) Nat Struct Mol Biol. 11 121-127.
  • Brodersen, D.E. et al. (2008) Biochim Biophys Acta. 1779, 532-537.
  • Wu, L., Fan J. and Belasco, J.G. (2006) Proc Natl Acad Sci U S A. 103, 4034-4039.
  • Izaurralde, E. et al. (2009) RNA 15, 21-32.
  • Martin, G. and Keller, W. (1998) RNA 4, 226-230.
  • Gauss-Müller, V. et al. (2001) Nucleic Acids Res. 29 E57-7
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Concentration1.25 U/μL
For Use With (Application)Poly(A) tail-length detection
Quantity1 kit
Shipping ConditionDry Ice
Starting MaterialTotal Eukaryotic RNA (100 ng to 2 μg)
PolymerasePoly(A) RNA Polymerase
Unit SizeEach
Contents & Storage
Sufficient reagents for 5 G/I tailing, 20 reverse transcription, and 80 PCR reactions.

HeLa Total RNA and Control Human Actin PCR Forward Primer to verify components and protocol.

Store reagents at -15°C to -30°C in a non-frost-free freezer.
Store HeLa Total RNA at -80°C.