Specifications
Boiling Point1390°C (1013 hPa)
CAS1310-73-2
Chemical Name or MaterialSodium hydroxide
ColorWhite
Melting Point323°C
Molecular FormulaNaOH
PackagingPolyethylene pail, screw top
SynonymSodium hydrate, Soda lye, Caustic soda, Lye
GradeFCC/NF
Molecular Weight (g/mol)40
Percent Purity95.0-100.5%
Quantity12 kg
Unit SizeEach