Product Image
Invitrogen™

Anti-Human Foxp3 Staining Set APC, eBioscience™

This Anti-Human Foxp3 Staining Set contains the buffers and monoclonal antibody necessary to successfully stain and identify Foxp3 cells. TheRead more
Have Questions?
Catalog NumberQuantity
77-5774-401 kit
Catalog number 77-5774-40
Price (EUR)
373,41
Online exclusive
461,00
Save 87,59 (19%)
Each
Quantity:
1 kit
Price (EUR)
373,41
Online exclusive
461,00
Save 87,59 (19%)
Each
This Anti-Human Foxp3 Staining Set contains the buffers and monoclonal antibody necessary to successfully stain and identify Foxp3 cells. The 236A/E7 monoclonal antibody reacts with human Foxp3 also known as FORKHEAD BOX P3, SCURFIN, and JM2. Foxp3 is a 49-55 kDa protein and a member of the forkhead/winged-helix family of transcription factors. It was identified as the gene responsible for the X-linked lymphoproliferative disease observed in scurfy (sf) mice and in the human disorder, X-linked autoimmunity-allergic dysregulation syndrome (XLAAD). Constitutive expression of Foxp3 mRNA has been shown in CD4+CD25+ regulatory T cells (Treg), and ectopic expression of Foxp3 in CD4+CD25- cells imparts a Treg phenotype in these cells.

The 236A/E7 antibody has been shown to crossreact with rhesus macaque, sooty mangabey, and cynomolgus Foxp3 236A/E7 recognizes a different epitope of Foxp3 than PCH101 and 150D/E4.

Isotype
IgG1

Laser
Red Laser

Emit
660 nm

Excite
633 - 647 nm

Reported Application
Intracellular Staining Followed by Flow Cytometric Analysis

For Research Use Only. Not for use in diagnostic procedures.
Specifications
AntigenFoxp3
Clone236A/E7
Gene AliasForkhead Box P3, Scurfin, JM2
Primary or SecondaryPrimary
Product LineeBioscience
ApplicationsFlow Cytometry
ClassificationMonoclonal
ConjugateAPC
Host SpeciesMouse
IsotypeIgG1
Quantity1 kit
Regulatory StatusRUO
Target SpeciesCynomolgus, Human, Macaque, Monkey, Non-human Primate, Rhesus Monkey
Unit SizeEach
Contents & Storage
Store at 2-8°C. Light-sensitive material. Use within 6 months of opening or by date indicated on the bottle.