Regulatory requirements vary by country; products may not be available in your geographic area.

Recombinant Bovine PrP (lyophilized)

Catalog number: 7700001

 Related applications: Animal Health

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The amino acid sequence of this highly purified protein (rec bovPrP) corresponds to mature bovine PrP (amino acids 25-242). The protein is expressed in an E. coli K12 strain. Expressed rec bovPrP was solubilized in 6 M guanidine hydrochloride from inclusion bodies, oxidized and purified by various chromatographic steps. Its molecular weight is 23,686 Da. Rec BovPrP is ideal as standard and positive control for Western blot and ELISA.
For Veterinary Use Only.

Specifications

Product Line: Prionics™
Product Size: 100 µg
Research Category: Neurobiology
Species: Cattle⁄Bovine
Validated Application: Assay Standard, ELISA, Gel Electrophoresis, Western Blot

Documents

Manuals & protocols