Product Image
Lorne Laboratories

Anti-S Monoclonal

Lorne Anti-S monoclonal reagent will cause indirect agglutination (clumping) of test red cells, that carry the corresponding specific antigen, in the antiglobulin phase of testing. No agglutination generally indicates the absence of the corresponding specific antigen.
Have Questions?
Catalog NumberQuantity
7700021 x 2 mL
Catalog number 770002
Price (MXN)
-
Quantity:
1 x 2 mL
Anti-S is part of the MNS system and has been implicated in Haemolytic Transfusion Reactions and Haemolytic Disease in newborns.
  • Lorne Monoclonal Anti-S blood grouping reagent contains human monoclonal antibodies diluted in a phosphate buffer containing sodium chloride and bovine albumin
  • It is supplied at optimal dilution for use with indirect Coombs tube test & gel card techniques
  • Reagent vials should be stored at 2 - 8°C
  • Prolonged storage at temperatures outside this range may result in accelerated loss of reagent reactivity
  • Product is available in 2 mL with 24 months shelf life
Specifications
Quantity1 x 2 mL
Shelf Life24 Months
TypeReagent
Detectable AnalytesS Antigen
Unit SizeEach
Contents & Storage
2°C to 8°C