Sodium chloride, 99.0-100.5% (dried basis), granular, GenAR™, USP for haemodialysis, GMP, Macron Fine Chemicals™
Avantor™

Sodium chloride, 99.0-100.5% (dried basis), granular, GenAR™, USP for haemodialysis, GMP, Macron Fine Chemicals™

This product is GMP Manufactured and meets JP, FCC, BP, USP, and EP Requirements.
Have Questions?
Cambiar vistabuttonViewtableView
Cantidad:
500 g
2.5 kg
12 kg
110 lb.
220 lb.
10 x 10 kg
Envase:
Ver todo
Frasco de plástico/poli Mall polystormor
Número de catálogo 7713-04
Precio (CLP)
-
Cantidad:
500 g
Envase:
Frasco de plástico/poli Mall polystormor
This product is GMP Manufactured and meets JP, FCC, BP, USP, and EP Requirements.
This product is GMP Manufactured and meets JP, FCC, BP, USP, and EP Requirements.
Especificaciones
Punto de ebullición1413°C (1013 hPa)
CAS7647-14-5
Nombre del producto químico o materialSodium chloride
ColorBlanco
Punto de fusión801°C
Fórmula molecularNaCl
EnvaseFrasco de plástico/poli Mall polystormor
SinónimoSalt, Halite, Sea salt, Rock salt, Saline
GradoUSP
Peso molecular (g/mol)58.44
Porcentaje de pureza99.0-100.5%
Cantidad500 g
Unit SizeEach