Sodium chloride, 99.0-100.5% (dried basis), granular, GenAR™, USP for haemodialysis, GMP, Macron Fine Chemicals™
Avantor™

Sodium chloride, 99.0-100.5% (dried basis), granular, GenAR™, USP for haemodialysis, GMP, Macron Fine Chemicals™

This product is GMP Manufactured and meets JP, FCC, BP, USP, and EP Requirements.
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
2.5 kg
12 kg
110 lb.
220 lb.
10 x 10 kg
Packaging:
View all
Poly bottle
Catalog number 7713-06
Price (MXN)
-
Quantity:
2.5 kg
Packaging:
Poly bottle
This product is GMP Manufactured and meets JP, FCC, BP, USP, and EP Requirements.
This product is GMP Manufactured and meets JP, FCC, BP, USP, and EP Requirements.
Specifications
Boiling Point1413°C (1013 hPa)
CAS7647-14-5
Chemical Name or MaterialSodium chloride
ColorWhite
Melting Point801°C
Molecular FormulaNaCl
PackagingPoly bottle
SynonymSalt, Halite, Sea salt, Rock salt, Saline
GradeUSP
Molecular Weight (g/mol)58.44
Percent Purity99.0-100.5%
Quantity2.5 kg
Unit SizeEach