Sodium chloride, 99.0-100.5% (dried basis), granular, GenAR™, USP for haemodialysis, GMP, Macron Fine Chemicals™
Avantor™

Sodium chloride, 99.0-100.5% (dried basis), granular, GenAR™, USP for haemodialysis, GMP, Macron Fine Chemicals™

This product is GMP Manufactured and meets JP, FCC, BP, USP, and EP Requirements.
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
2.5 kg
12 kg
110 lb.
220 lb.
10 x 10 kg
Packaging:
View all
Polyethylene drum, open head
Drum, HDPE Flowmor™ package
Catalog number 7713-25
Price (MXN)
-
Quantity:
110 lb.
Packaging:
Polyethylene drum, open head
This product is GMP Manufactured and meets JP, FCC, BP, USP, and EP Requirements.
This product is GMP Manufactured and meets JP, FCC, BP, USP, and EP Requirements.
Specifications
Boiling Point1413°C (1013 hPa)
CAS7647-14-5
Chemical Name or MaterialSodium chloride
ColorWhite
Melting Point801°C
Molecular FormulaNaCl
PackagingPolyethylene drum, open head
SynonymHalite, Sea salt, Rock salt, Saline
GradeUSP
Molecular Weight (g/mol)58.44
Percent Purity99.0-100.5%
Quantity110 lb.
Unit SizeEach