Product Image
Thermo Scientific™
Recording Balance D
The Recording Balance D has been discontinued, however we continue to provide services and support for this product.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberModel
7730050Recording Balance D-110
773-0020Recording Balance D-101
773-0040Recording Balance D-200
7730010Recording Balance D-100
Catalog number 7730050
Price (KRW)
-
Model:
Recording Balance D-110
The Recording Balance D has been discontinued, however we continue to provide services and support for this product.
Please see below for available services and support for the Recording Balance D.
This product is no longer being manufactured. We remain committed to providing you with ongoing access to services and support for the Recording Balance D.
As of December 31, 2010, the items below are no longer available.
Specifications
DescriptionRecording Balance D-202; No Vacuum; Max capacity: 1.5 / 3.5gr; max sensitivity: 0.1ug / 1ug
ModelRecording Balance D-110
RepeatabilityNo load 50% lift: 100μg; Fraction of load, 50% lift: 0.002%
Weight (Metric)100 g
Unit Size-