cAMP-dependent protein kinase (PKA) inhibitor. This peptide is a highly specific inhibitor of cAMP-dependent protein kinase (PKA).Format/Formulation: Lyophilized trifluoroacetate salt,Read more
Have Questions?
Catalog Number
Quantity
77409
1 mg
Catalog number 77409
Price (BRL)
-
Quantity:
1 mg
cAMP-dependent protein kinase (PKA) inhibitor. This peptide is a highly specific inhibitor of cAMP-dependent protein kinase (PKA).
Format/Formulation: Lyophilized trifluoroacetate salt, >97% purity by HPLC.
Molecular Weight: 1208
For Research Use Only. Not for use in diagnostic procedures.