Thermo Scientific™
Shandon™ Finesse™ Fine Orienting Head
Related applications:
Anatomical Pathology
Designed for the Thermo Scientific Shandon™ Finesse ME/E Series only, the Thermo Scientific™ Finesse Fine Orienting Head is of excellent quality.
Back to top
  • Allows x-y orientation of the specimen
  • Supplied with the Finesse ME+ and E+ model microtomes only