Thermo Scientific™

Pierce™ Recombinant Protein L

Catalog number: 77680
Thermo Scientific™

Pierce™ Recombinant Protein L

Catalog number: 77680
Catalog Number
77680
Unit Size
500 mg
Price (USD)
Price: 2,985.00
Your Price:
Availability
-
Quantity
Catalog Number
Unit Size
1 mg
Price (USD)
Price: 115.00
Your Price:
Availability
-
Quantity
Catalog Number
Unit Size
50 mg
Price (USD)
Price: 816.00
Your Price:
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
77680500 mg
Price: 2,985.00
Your Price:
-
211891 mg
Price: 115.00
Your Price:
-
7767950 mg
Price: 816.00
Your Price:
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Purified (unconjugated) Thermo Scientific Pierce Recombinant Protein L is useful as the basis for preparing various kinds of probes or affinity media for detection or purification of mouse and human antibodies in immunoassays and antibody purification protocols.

Features of Recombinant Protein L:

• Contains four immunoglobulin-binding domains per protein
• Binds members of all classes of Ig (IgG, IgM, IgA, IgE and IgD) that contain kappa light chains
• Binds single chain variable fragments (scFv) and Fab fragments that contain kappa light chains

Protein L is an immunoglobulin-binding protein that was originally derived from the bacteria Peptostreptococcus magnus, but is now produced recombinantly. Protein L has the unique ability to bind through kappa light chain interactions without interfering with an antibody's antigen-binding site. This gives Protein L the ability to bind a wider range of Ig classes and subclasses than other antibody-binding proteins. Protein L will bind to all classes of Ig (IgG, IgM, IgA, IgE and IgD). Protein L will also bind single chain variable fragments (scFv) and Fab fragments. Protein L binds kappa I, III, and IV in human and kappa I on mouse.

Properties of Recombinant Protein L:
• Source: E. coli
• Molecular Weight: ∼35,800 (Apparent MW by SDS-PAGE: 36,000)
• Form: salt-free powder
• A280 of 0.1% solution: ∼0.80
• Isoelectric point (pI): 4.8

Related Products
Pierce™ Recombinant Protein L, Biotinylated
Pierce™ Recombinant Protein L, Peroxidase Conjugated
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Quantity
500 mg
Product Line
Pierce™
Form
Powder
Protein Subtype
Protein L
Target
Antibodies

Contents & Storage

Upon receipt store at 4°C.

Note: Store reconstituted product for up to one month at 4°C. For long-term storage, aliquot, add an equal volume of glycerol and store at -20°C.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text