Trisodium citrate, dihydrate, 99.0-100.5% (by anhydrous basis), crystals, GenAR™, USP, Multi-Compendial, GMP, Macron Fine Chemicals™
Avantor™

Trisodium citrate, dihydrate, 99.0-100.5% (by anhydrous basis), crystals, GenAR™, USP, Multi-Compendial, GMP, Macron Fine Chemicals™

This product is GMP Manufactured and meets JP, BP, USP, and EP Requirements.
Have Questions?
Cambiar vistabuttonViewtableView
Cantidad:
500 g
2.5 kg
12 kg
100 lb.
Número de catálogo 7773-06
Precio (MXN)
-
Cantidad:
2.5 kg
This product is GMP Manufactured and meets JP, BP, USP, and EP Requirements.
Especificaciones
CAS4-3-6132
Nombre del producto químico o materialTrisodium citrate dihydrate
ColorBlanco
Punto de fusión150°C
Fórmula molecularHOC(COONa)(CH2COONa)2·2H2O
EnvaseAmber glass bottle
SinónimoCitric acid tridosium salt dihydrate, Sodium citrate dihydrate, Sodium citrate tribasic dihydrate, Trisodium citrate
GradoMulti-Compendial
Peso molecular (g/mol)294.1
Porcentaje de pureza99.0-100.5%
Cantidad2.5 kg
Unit SizeEach