ABC SSP UniTray® Kit
Catalog number: 7800010
Invitrogen™  Related applications: Diagnostic Testing
Error loading your content!
  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons
Your on-site stock
›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top
Description
•Provides low to intermediate HLA typing results
•Products offered in a variety of formats, including combo kits
•Quick turn-around-time makes this technology ideal for on-call typing
• Pre-dispensed liquid primer mixes
•Includes pre-aliquotted amplification buffer
•Compatible with UniMatch Plus Analysis software
•Allele-updating availabe via the web
Contents & storage
Store in freezer (-5 to -30°C).
Documents
Manuals & protocols
Recommended products