ExoSAP-IT™ PCR Product Cleanup
ExoSAP-IT™ PCR Product Cleanup
Applied Biosystems™
ExoSAP-IT™ PCR Product Cleanup
Features and Benefits of the ExoSAP-IT™ PCR Product Cleanup• Conserves PCR samples — 100% recovery of both short and longLeia mais
Have Questions?
Número do catálogoQuantity
78250.40.UL40 μL
Número do catálogo 78250.40.UL
Preço (BRL)
315,76
Each
Adicionar ao carrinho
Quantity:
40 μL
Preço (BRL)
315,76
Each
Adicionar ao carrinho
Features and Benefits of the ExoSAP-IT™ PCR Product Cleanup
Conserves PCR samples — 100% recovery of both short and long PCR products
One tube/one step PCR cleanup — Add ExoSAP-IT reagent directly to PCR product
Eliminates spin columns — Decreases time and expense while increasing yield
Removes contaminating primers and dNTPs — No interference in downstream applications
Scalable — Economical for high-throughput purification
Simple processing — Robotic-friendly; Replaces beads, filtrations, and plates
Generates less waste than columns
ExoSAP-IT reagent is designed for simple, quick PCR cleanup for downstream applications, such as DNA sequencing or Single Nucleotide Polymorphism (SNP) analysis. When PCR amplification is complete, any unconsumed dNTPs and primers remaining in the PCR product mixture will interfere with these methods. ExoSAP-IT removes these contaminants.
ExoSAP-IT is added directly to the PCR product and incubated at 37°C for 15 minutes (Fig. 1). After PCR treatment, ExoSAP-IT is inactivated simply by heating to 80°C for 15 minutes.
ExoSAP-IT single-step PCR cleanup utilizes two hydrolytic enzymes, Exonuclease I and Shrimp Alkaline Phosphatase (SAP), together in a specially formulated buffer, to remove unwanted dNTPs and primers from PCR products. Exonuclease I removes residual single-stranded primers and any extraneous single-stranded DNA produced in the PCR. SAP removes the remaining dNTPs from the PCR mixture.
Rapid PCR Product Cleanup Protocol
ExoSAP-IT requires only one pipetting step and two incubations. Just add ExoSAP-IT to the PCR product and within 30 minutes sequencing or SNP analysis can be performed.
Simple: Single-Step
The method is designed to require a minimum of ‘hands-on’ time. Enzymatic removal of excess primers and unincorporated nucleotides occurs in one easy step by using ExoSAP-IT reagent in a single tube or microtiter well. Only simple pipette transfers are required, therefore, many samples can be processed at once, either manually or with robotics.
No Sample Loss
Use of ExoSAP-IT reagent eliminates all gel or column purifications, sedimentations, filtrations, beads, and/or magnetic separations. There is 100% recovery of both short and long PCR products with ExoSAP-IT (Fig. 2).
Achieve High Data Quality from PCR Products
ExoSAP-IT reagent may be used as an effective cleanup method prior to fluorescent or radioactive DNA sequencing, SNP analysis, or any other application requiring a PCR product free of excess nucleotides and primers.
References:
DUGAN, K. A., LAWRENCE, H. S., HARES, D. R., FISHER, C. L. AND BUDOWLE B. (2002) J. Forensic Sci 47, 811-818.
HANKE, M. AND WINK, M. (1994) BioTechniques 17, 858-860.
MU, J., DUAN, J., MAKOVA, K., JOY, D., HUYNH, C., BRANCH, O., LI, W. AND SU, X. (2002) Nature 418, 323-326.
SILVA, JR., W. A., COSTA, M. C. R., VALENTE, V., DE FREITAS SOUSA, J., PACÓ-LARSON, M. L., ESPREAFICO, E. M., CAMARGO, S. S., MONTEIRO, E., DE JESUS, A., HOLANDA, M. A., ZAGO, M. A., SIMPSON, A. J. G. AND NETO, E. D. (2001) BioTechniques 30, 537-542.
WERLE, E., SCNEIDER C., RENNER, M., VÖLKER, M. AND FIEHN, W. (1994) Nucleic Acids Res. 22, 4354-4355.
For Research Use Only. Not for use in diagnostic procedures.
Especificações
DescriptionExoSAP-IT™ PCR Product Clean-up
For Use With (Application)PCR Purification
Quantity40 μL
Test Time15 min
FormatSpin Column
Unit SizeEach
Frequently asked questions (FAQs)
Can ExoSAP-IT PCR cleanup reagents be used with automatic handling systems?
The HT ExoSAP-IT Fast High-Throughput PCR Product Cleanup reagent is designed for automated handlers as it has a lower viscosity than standard ExoSAP-IT PCR Product Cleanup reagent, which contains some glycerol.
After purification with ExoSAP-IT PCR cleanup reagents, what is the recommended quantification method?
The Quant-iT PicoGreen dsDNA Assay Kit is recommended for quantifying PCR products that have been cleaned with ExoSAP-IT reagent. ExoSAP-IT PCR cleanup results in 100% recovery regardless of amplicon length, and if the PCR product was quantified prior to ExoSAP-IT cleanup, additional quantification is not necessary. We do not recommend using a spectrophotometer as degraded products will be included in the quantification.
Can ExoSAP-IT PCR cleanup reagents be used for removal of primers after first-strand cDNA synthesis?
For use with reverse transcription, ExoSAP-IT does not degrade the DNA-RNA hybrid. However, it will degrade single-stranded cDNA if a 3' -OH end is present.
Will the two tracking dyes used for visualization in the DreamTaq Hot Start Green PCR Master Mix interfere with the ExoSAP-IT PCR cleanup reagents?
The tracking dyes used in the DreamTaq Hot Start Green PCR Master Mix have not been shown to interfere with functionality of the ExoSAP-IT cleanup reagents.
Can ExoSAP-IT PCR cleanup reagents be used to cleanup PCR products amplified using High Fidelity (HiFi) DNA polymerases?
The proofreading function of HiFi DNA polymerases can cause partial degradation of PCR products during extended incubation at 80°C. ExoSAP-IT Express reagent is recommended for HiFi PCR cleanup because it only requires 1 minute at 80°C for inactivation.