Halt™ Phosphatase Inhibitor Cocktail

Catalog number: 78420B

Thermo Scientific™  Related applications: Protein Purification & Isolation

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Thermo Scientific Halt Phosphatase Inhibitor Cocktail preserves the phosphorylation state of proteins during and after cell lysis or tissue protein extraction.

Features of Halt Phosphatase Inhibitor Cocktail:

Devoid of protease inhibitors—To protect against both proteases and phosphatases, use Halt Protease and Phosphatase Inhibitor Cocktail.
Dialyzable— Either dialyze or desalt sample to effectively remove inhibitors from cell extracts before performing procedures that are discovered to be incompatible with cocktail components.
Supplied as a 100X concentrate—effective when used at a final concentration of 1X; however, samples that contain particularly high levels of phosphatase activity may require a final concentration of 2-3X.
Compatibility— compatible with most detergent-based cell lysis reagents, including all Thermo Scientific Pierce Cell Lysis Solutions.
All-in-one format—inhibits both serine-threonine phosphatases and protein tyrosine phosphatases
Compatiblewith popular protein assays—works with standard protein assays, including BCA and coomassie (Bradford) assays
Convenient storage—refrigerator (2 to 8°C) storage saves valuable space in the freezer and provides for immediate use
Multiple sizes—select a package size of the 100X cocktail that is ideal for any particular scale and workflow of protein extraction experiment

Halt Phosphatase Inhibitor Cocktail protects phosphoproteins from serine/threonine phosphatases as well as protein tyrosine phosphatases (PTPs) during cell lysis. The cocktail contains a mixture of four inhibitors of broad specificity, including sodium fluoride, sodium orthovanadate, sodium pyrophosphate and beta-glycerophosphate. Conveniently packaged in a 100X format, the cocktail is stable at 4°C and is suitable for use with any cell or tissue extract. The cocktail preserves protein phosphorylation in immunoprecipitation and kinase assays.

Phosphorylation and dephosphorylation comprise a molecular on/off switch that regulates many key biological pathways within the cell, including signal transduction, cell division and apoptosis. To effectively study phosphorylation activation states, one must inactivate phosphatases to prevent their uncontrolled activity upon cell lysis. Halt Phosphatase Inhibitor Cocktail accomplishes this task.

Related Products
Pierce™ Phosphatase Inhibitor Mini Tablets
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Reagent Type: Phosphatase Inhibitor
Form: Liquid
Product Line: Halt™
Product Size: Custom

Contents & storage

Upon receipt store product at 4°C. Do not freeze.

Documents

Manuals & protocols