Licensed UniCheck® Kit, includes Taq DNA Polymerase
Catalog number: 7891110
 Related applications: Diagnostic Testing
Error loading your content!
  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons
Your on-site stock
›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top
Description
Uses SSP technology and the standard UniTray protocol. A single primer mix is plated in all 96 wells of a polycarbonate tray. A positive DNA control is included for testing all 96 wells. UniCheck contains primer pairs that can detect variations in both denaturing and annealing temperatures and will not amplify under sub-optimal conditions.
Contents & storage
Store in freezer (-5 to -30°C).
Documents
Manuals & protocols