Cytokine Genotyping Kit
Catalog number: 7892010
Invitrogen™  Related applications: Diagnostic Testing
Error loading your content!
  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons
Your on-site stock
›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top
Description
Cytokines show a low degree of polymorphism, usually restricted to the promoter region of the cytokine genes. Several authors describe that these polymorphisms have both functional and clinical relevance. Levels of various cytokines have been found to influence graft outcome.
The Pel-Freez Cytokine Genotyping Kit identifies polymorphisms of IL1a, IL1b, IL1R, IL1RA, IL4Ra, IL12, yIFN, TGFb, TNFa, IL2, IL4, IL6, and IL10 using a standard SSP technique. Each tray contains forty-eight formulations of specific primer sets used to amplify genomic DNA. Two tests contained on each tray. Primer sets supplied lyophilized and include PCR buffer.
Contents & storage
Store in freezer (-5 to -30°C).
Documents
Manuals & protocols