Licensed DNA Wipe Test, includes Taq DNA Polymerase
Catalog number: 789981
 Related applications: Diagnostic Testing
Error loading your content!
  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons
Your on-site stock
›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top
Description
Single master primer mix to identify Class I and Class II contamination. UniTray DNA Wipe Test covers specificities of Class I A, B, and C and Class II DR and DQ loci. In a convenient pre-aliquoted tray. Contamination test is designed for use with the SSP UniTray kits. Used for the detection of genomic DNA, cloned DNA, and amplified PCR product.
Contents & storage
Store in freezer (-5 to -30°C).
Documents
Manuals & protocols