Orion 8000 Sample Sequencer

Orion 8000 Sample Sequencer

Confidently perform silica analysis with the Thermo Scientific™ Orion™ 8030cX Silica Analyzer. Designed to provide accurate, effortless water analysis, the Orion 8030cX specializes in online measurement of reactive silica.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberSample Streams
8000SEQ44
8000SEQ22
8000SEQ66
Catalog number 8000SEQ4
Price (EUR)
-
Sample Streams:
4
Confidently perform silica analysis with the Thermo Scientific™ Orion™ 8030cX Silica Analyzer. Designed to provide accurate, effortless water analysis, the Orion 8030cX specializes in online measurement of reactive silica. Powered by advanced colorimetric technology, the Orion 8030cX delivers premium precision, greater productivity and enhanced durability. Supporting ultrapure and process water applications, the Orion 8030cX conveniently delivers precise, reliable measurement you can depend on. Unleash the possibilities and conquer the freedom of fast, confident decision making.

Confident Results

  • Low Detection Limit (0.5 ppb)
  • High Accuracy (≤1 ppb or 1% for range 0-500 ppb)
  • Patented Fluid Delivery & Quantification System
  • Reduced Phosphate Interferences

Low-Cost Ownership

  • 100 Days of Continuous Run Time (w/ 2L reagent and 15-minute measurement frequency)
  • Low Reagent Consumption (200 μL per measurement)
  • Separate Chemical Waste Collection (decreased disposal expenses)

Easy to Use

  • Automated Calibration
  • Automated Validation & Verification
  • Color Touchscreen with Intuitive Interface

Maximum Uptime

  • Automated Cleaning
  • Scheduled Measurements
  • Alert Notification System
  • Alarm System w/ Troubleshooting Guide
  • IP65 Certified (splash/ dust proof)

Markets

  • Silica monitoring for ultrapure water, demineralized water, deionized water, boiler water and steam condensate in:
  • Power Generation
  • Oil & Gas
  • Chemical Production
  • Semiconductor Manufacturing
  • Pulp & Paper

Applications

  • Monitoring and controlling silica levels in ultrapure and industrial process water to:
  • Prevent buildup on turbine blades, heat exchangers, boilers and more
  • Reduce costly and time-consuming cleaning procedures
  • Identify reverse osmosis (RO) breakdown and saturation of ion exchange resins
  • Maximize thermal efficiencies of steam passageways
  • Safeguard the health of critical plant assets by reducing potential for turbine vibration
Specifications
Accuracy≤1 ppb or 1% for range 0-500 ppb
Alarm OutputAlarm System w/ Troubleshooting Guide
CalibrationAutomated Calibration
Certifications/ComplianceIP65 Certified (splash/ dust proof)
DescriptionThe optional Thermo Scientific™ Orion™ 8000 Series SEQ4 Sample Sequencer enables monitoring of up to four (4) sample streams from various locations without the need for additional instrumentation.
Detection Limits0.5 ppb
Display TypeColor Touchscreen with Intuitive Interface
For Use With (Application)
  • Monitoring and controlling silica levels in ultrapure and industrial process water to:
  • Prevent buildup on turbine blades, heat exchangers, boilers and more
  • Reduce costly and time-consuming cleaning procedures
  • Identify reverse osmosis (RO) breakdown and saturation of ion exchange resins
  • Maximize thermal efficiencies of steam passageways
  • Safeguard the health of critical plant assets by reducing potential for turbine vibration
Sample Streams4
TypeSample Sequencer
Unit SizeEach