Regulatory requirements vary by country; products may not be available in your geographic area.
TypiFix™ High Throughput Extractor EA-TE2
Catalog number: 8001630
Error loading your content!
  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons
Your on-site stock
›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top
Description
Fully automated extractor for the retrieval of tissue samples from TypiFix™ sample containers. Four sample racks are included. Ideal for high throughput. The TE2 extractor can process >500 samples per hour.
For Veterinary Use Only.
Specifications
Product Line: Prionics™
Product Size: 1 piece
Type: Other Accessories
Documents
Manuals & protocols