Regulatory requirements vary by country; products may not be available in your geographic area.
TypiFix™ Cattle Ear Tag HH, blank, yellow
Catalog number: 8003224
Error loading your content!
  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons
Your on-site stock
›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top
Description
The innovative TypiFix™ system combines conventional ear tagging with tissue sampling in one simple step. During the process of tagging livestock, this sampling technology automatically collects and seals tissue samples in TypiFix™ sample containers, conserving proteins and nucleic acids. The system can therefore be used for simultaneous sampling for BVDV and genotyping.
TypiFix™ sample containers mummify samples by airtight locking of samples together with the desiccant. Thanks to this proprietary conservation process, the samples can be stored at room temperature without any deterioration. Sample containers can be easily transported to the laboratory by regular mail, and require no cooling.
The system is compliant with EU regulations on tamperproof animal identification. It is an efficient and cost-effective solution for identifying and tracking animals and includes processes for automation and data management.
This unlabeled ear tag was developed for use with cattle. The tags can be labeled individually by the user.
For Veterinary Use Only.
Specifications
Product Line: Prionics™
Product Size: 1 piece
Type: Other Accessories
Documents
Manuals & protocols