Thermo Scientfic™ Orion™ 8010cX Process Ammonia Analyzer
El producto real puede variar
Thermo Scientfic™ Orion™ 8010cX Process Ammonia Analyzer
Thermo Scientfic™ Orion™ 8010cX Process Ammonia Analyzer
Thermo Scientfic™ Orion™ 8010cX Process Ammonia Analyzer
Thermo Scientific™
Thermo Scientfic™ Orion™ 8010cX Process Ammonia Analyzer
Thermo Scientific™ Orion™ 8010cX is designed to provide easy to use, accurate, reliable and low maintenance ammonia online monitoring solution.
Have Questions?
Cambiar vistabuttonViewtableView
Número de catálogoTipoCantidad
8010MKKit de mantenimiento1 kit
8010002Solución estándar de amoníaco2 mg/L
8010005Solución estándar de amoníaco5 mg/L
8010030Solución estándar de amoníaco30 mg/L
8010100Solución estándar de amoníaco100 mg/L
8010500Solución estándar de amoníaco500 mg/L
8010REKit de reactivo de amoníaco1 kit
8010RFG115Refrigerador (115 V)1 unidad
8010RFG220Refrigerador (220 V)1 unidad
8010CXAnalizador de amoníaco1 unidad
Número de catálogo 8010MK
Precio (MXN)
-
Tipo:
Kit de mantenimiento
Cantidad:
1 kit
Thermo Scientific™ Orion™ 8010cX is designed to provide easy to use, accurate, reliable and low maintenance ammonia online monitoring solution.8010cX is based on standard salicylic acid method, method traceable and reagents low toxicity. Professional industrial design, user interface design, modularity design, software function design, anti-interference measurement process design and algorithm enabled 8010cX can be applied in various different applications, to meet regulatory compliance and process control requirements.
Thermo Scientific™ Orion™8010cX is:
  • Designed for accurate and reliable online measurement, meet waste water discharge regulatory requirements and process control requirements.
  • Automated ranging for validity accurate determination of ammonia
  • Feature-rich software and intuitive and graphical user interface
  • Special measurement process design and algorithm enabled analyzer have good anti-interference (color and turbidity) capability.
  • Compact size, small installation footprint
  • Modularity design, IP65 protection level and long lifetime components make sure long-term operation.
  • Low operation cost and maintenance cost – low reagent consumption and low chemical waste generation.
Especificaciones
Cantidad1 kit
TipoKit de mantenimiento
Unit SizeEach