Thermo Scientfic™ Orion™ 8010cX Process Ammonia Analyzer
Actual product may vary
Thermo Scientfic™ Orion™ 8010cX Process Ammonia Analyzer
Thermo Scientfic™ Orion™ 8010cX Process Ammonia Analyzer
Thermo Scientfic™ Orion™ 8010cX Process Ammonia Analyzer
Thermo Scientific™

Thermo Scientfic™ Orion™ 8010cX Process Ammonia Analyzer

Thermo Scientific™ Orion™ 8010cX is designed to provide easy to use, accurate, reliable and low maintenance ammonia online monitoring solution.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberTypeQuantity
8010RFG220Refrigerator (220V)1 Ea.
8010002Ammonia Standard Solution2 mg/L
8010005Ammonia Standard Solution5 mg/L
8010030Ammonia Standard Solution30 mg/L
8010100Ammonia Standard Solution100 mg/L
8010500Ammonia Standard Solution500 mg/L
8010MKMaintenance Kit1 Kit
8010REAmmonia Reagent Kit1 Kit
8010RFG115Refrigerator (115 V)1 Ea.
8010CXAmmonia Analyzer1 Ea.
Catalog number 8010RFG220
Price (USD)
-
Type:
Refrigerator (220V)
Quantity:
1 Ea.
Thermo Scientific™ Orion™ 8010cX is designed to provide easy to use, accurate, reliable and low maintenance ammonia online monitoring solution.8010cX is based on standard salicylic acid method, method traceable and reagents low toxicity. Professional industrial design, user interface design, modularity design, software function design, anti-interference measurement process design and algorithm enabled 8010cX can be applied in various different applications, to meet regulatory compliance and process control requirements.

Thermo Scientific™ Orion™8010cX is:

  • Designed for accurate and reliable online measurement, meet waste water discharge regulatory requirements and process control requirements.
  • Automated ranging for validity accurate determination of ammonia
  • Feature-rich software and intuitive and graphical user interface
  • Special measurement process design and algorithm enabled analyzer have good anti-interference (color and turbidity) capability.
  • Compact size, small installation footprint
  • Modularity design, IP65 protection level and long lifetime components make sure long-term operation.
  • Low operation cost and maintenance cost – low reagent consumption and low chemical waste generation.
Specifications
For Use With (Equipment)Reagent 2
Quantity1 Ea.
TypeRefrigerator (220V)
Unit SizeEach