Portable Table
Fisher Scientific™
Portable Table
Allows safe and easy transport of equipment from countertop to cart.
Have Questions?
카탈로그 번호 8025000
LABCONCO-8025000-EACH으로도 사용됨
제품 가격(KRW)
-
Allows safe and easy transport of equipment from countertop to cart.
  • Powder-coated 18-gauge 1 in. tubular steel frame
  • Safely transports heavy instruments and bulky equipment
  • Two open shelves
  • Glacier white frame with gray accents
  • Handle bar on each end with ergonomic extruded rubber grip
  • 4 in. diameter casters, two with toe locks
  • Supports loads up to 400 pounds
  • ID tag
  • Lifetime guarantee
  • Ready-to-roll, full factory assembly
사양
치수(길이 x 폭 x 높이)34.88 x 19.0 x 36.38 in. (88.6 x 48.3 x 92.4 cm)
포함Two open shelves; 4 in. diameter casters, two with toe locks; Extruded rubber handle grips; Glacier white, epoxy-coated 18 gauge welded tubular steel frame; ID tag
캐스터 수4
하중 지지 용량(영국식 단위)400 lb.
하중 지지 용량(미터법)181 kg
물질Steel
선반 수2
제품 유형Equipment Cart
Unit SizeEach