Sodium sulfate, anhydrous, ≥99.0% (dry basis), USP, GMP, Macron Fine Chemicals™
Avantor™

Sodium sulfate, anhydrous, ≥99.0% (dry basis), USP, GMP, Macron Fine Chemicals™

GMP Manufactured Product. Meets U.S.P Requirements.
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
2.5 kg
12 kg
100 kg
350 lb.
Packaging:
View all
Round poly bottle
Catalog number 8028-06
Price (MXN)
-
Quantity:
2.5 kg
Packaging:
Round poly bottle
GMP Manufactured Product. Meets U.S.P Requirements.
GMP Manufactured Product. Meets U.S.P Requirements.
Specifications
Boiling Point1429°C (1013 hPa)
CAS7757-82-6
Chemical Name or MaterialSodium sulfate
ColorWhite
Melting Point888°C
Molecular FormulaNa2SO4
PackagingRound poly bottle
SynonymSulphuric acid disodium salt, Bisodium sulphate, Sulfuric acid disodium salt, Bisodium sulfate, Disodium sulphate
GradeUSP
Molecular Weight (g/mol)142.04
Percent Purity≥99.0%
Quantity2.5 kg
Unit SizeEach