Specifications
Boiling Point1429°C (1013 hPa)
CAS7757-82-6
Chemical Name or MaterialSodium sulfate
ColorWhite
Melting Point888°C
Molecular FormulaNa2SO4
PackagingMall polystormor poly/plastic bottle
SynonymSulphuric acid disodium salt, Bisodium sulphate, Sulfuric acid disodium salt, Bisodium sulfate, Disodium sulphate
GradeUSP
Molecular Weight (g/mol)142.04
Percent Purity≥99.0%
Quantity500 g
Unit SizeEach