Sodium sulfate, anhydrous, ≥99.0% (dry basis), USP, GMP, Macron Fine Chemicals™
Avantor™

Sodium sulfate, anhydrous, ≥99.0% (dry basis), USP, GMP, Macron Fine Chemicals™

GMP Manufactured Product. Meets U.S.P Requirements.
Have Questions?
Cambiar vistabuttonViewtableView
Cantidad:
500 g
2.5 kg
12 kg
100 kg
350 lb.
Envase:
Ver todo
Polyethylene pail, screw top
Mall stakmor case
Número de catálogo 8028-88
Precio (CLP)
-
Cantidad:
12 kg
Envase:
Polyethylene pail, screw top
GMP Manufactured Product. Meets U.S.P Requirements.
GMP Manufactured Product. Meets U.S.P Requirements.
Especificaciones
Punto de ebullición1429°C (1013 hPa)
CAS7757-82-6
Nombre del producto químico o materialSodium sulfate
ColorBlanco
Punto de fusión888°C
Fórmula molecularNa2SO4
EnvasePolyethylene pail, screw top
SinónimoSulphuric acid disodium salt, Bisodium sulphate, Sulfuric acid disodium salt, Bisodium sulfate, Disodium sulphate
GradoUSP
Peso molecular (g/mol)142.04
Porcentaje de pureza≥99.0%
Cantidad12 kg
Unit SizeEach