B·R·A·H·M·S™ uE3 KRYPTOR™
Thermo Scientific™
B·R·A·H·M·S™ uE3 KRYPTOR™
B·R·A·H·M·S provides a complete portfolio of biomarkers for the most reliable results in prenatal screening. Get the quadruple test for trisomy screening in second trimester of pregnancy with B·R·A·H·M·S uE3 KRYPTOR in less than 40 minutes.
Have Questions?
Ansicht ändernbuttonViewtableView
KatalognummerTyp
803075NImmunofluorescent Assay
080391NKalibrator
080392NKontrollen
Katalognummer 803075N
Preis (EUR)
-
Typ:
Immunofluorescent Assay
B·R·A·H·M·S provides a complete portfolio of biomarkers for the screening of chromosomal abnormalities in first and second trimester of pregnancy. B·R·A·H·M·S uE3 KRYPTOR can be used as an aid to provide an early identification of the pregnancies at risk of trisomy 21 in second trimester.
US Only: Product is not 510k cleared in US.
Get confidence in your results with the high precision of B·R·A·H·M·S uE3 KRYPTOR and the full quadruple test on B·R·A·H·M·S KRYPTOR GOLD.
  • Incubation time of only 19 minutes for a short turnaround time
  • Allows measurement in serum
  • Large capacity with 75 tests
  • Kit stable up to 15 days after opening onboard the instrument
  • Easy workflow with only 2 calibrators and one calibration every week
  • Fits your throughput need with B·R·A·H·M·S KRYPTOR compact PLUS or B·R·A·H·M·S KRYPTOR GOLD
Thermo Fisher Scientific products are distributed globally and their uses, applications, indications, claims and availability of products in each country depend on local regulatory marketing authorization status, please consult the Instructions For Use (IFU) available in your country.
Specifications
Analysezeit19 minutes
NachweisverfahrenImmunofluorescence
Zur Verwendung mit (Anwendung)For quantitative determination of unconjugated estriol (uE3). Risk assessment of fetal trisomy 21 in second trimester of pregnancy
EnthältKit for 75 determinations
ProbentypSerum
TypImmunofluorescent Assay
Nachweisbare AnalytenuE3
FormLyophilisiert
Unit SizeEach
Inhalt und Lagerung
Kit for 75 determinations, 2°C to 8°C