Orion™ 8030cX Silica Analyzer
Thermo Scientific™

Orion™ 8030cX Silica Analyzer

Confidently perform silica analysis with the Thermo Scientific™ Orion™ 8030cX Silica Analyzer. Designed to provide accurate, effortless water analysis, the Orion 8030cX specializes in online measurement of reactive silica.
Have Questions?
Ansicht ändernbuttonViewtableView
KatalognummerTypMenge
8030MKWartungs-Kit1 jeweils
7070FLTProbenvorfilter1 jeweils
803002Silika-Standardlösung20μ g/l
803020Silika-Standardlösung200 μg/l
803030Silika-Standardlösung1.000μ g/l
803040Silika-Standardlösung4000μ g/l
8030CLReinigungslösung1 jeweils
8030CXSilika-Analysator1 jeweils
8010PCMProfibus-Karte1 jeweils
8030REXSilika-Reagenzienkit1 jeweils
Katalognummer 8030MK
Preis (EUR)
-
Typ:
Wartungs-Kit
Menge:
1 jeweils
Confidently perform silica analysis with the Thermo Scientific™ Orion™ 8030cX Silica Analyzer. Designed to provide accurate, effortless water analysis, the Orion 8030cX specializes in online measurement of reactive silica. Powered by advanced colorimetric technology, the Orion 8030cX delivers premium precision, greater productivity and enhanced durability. Supporting ultrapure and process water applications, the Orion 8030cX conveniently delivers precise, reliable measurement you can depend on. Unleash the possibilities and conquer the freedom of fast, confident decision making.
Confident Results
  • Low Detection Limit (0.5 ppb)
  • High Accuracy (≤1 ppb or 1% for range 0-500 ppb)
  • Patented Fluid Delivery & Quantification System
  • Reduced Phosphate Interferences

 Low Cost Ownership
  • 100 Days of Continuous Run Time (w/ 2L reagent and 15-minute measurement frequency)
  • Low Reagent Consumption (200 μL per measurement)
  • Separate Chemical Waste Collection (decreased disposal expenses)

 Easy to Use
  • Automated Calibration
  • Automated Validation & Verification
  • Color Touchscreen with Intuitive Interface

 Maximum Uptime
  • Automated Cleaning
  • Scheduled Measurements
  • Alert Notification System
  • Alarm System w/ Troubleshooting Guide
  • IP65 Certified (splash/ dust proof)
Markets
Silica monitoring for ultrapure water, demineralized water, deionized water, boiler water and steam condensate in:
  • Power Generation
  • Oil & Gas
  • Chemical Production
  • Semiconductor Manufacturing
  • Pulp & Paper


Applications

Monitoring and controlling silica levels in ultrapure and industrial process water to:
  • Prevent buildup on turbine blades, heat exchangers, boilers and more
  • Reduce costly and time-consuming cleaning procedures
  • Identify reverse osmosis (RO) breakdown and saturation of ion exchange resins
  • Maximize thermal efficiencies of steam passageways
  • Safeguard the health of critical plant assets by reducing potential for turbine vibration
Specifications
BeschreibungThe Thermo Scientific™ Orion™ 8030cX Maintenance Kit includes replacement components for one (1) set of fluidic tubing and tube connection fittings for maintenance.
Zur Verwendung mit (Geräte)Orion 8030cX Silica Analyzer
Menge1 jeweils
TypWartungs-Kit
Unit SizePack of